siRNA Details
| Gene Symbol: Spag8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173555.1 | NP_001167026.1 | 7 | 1405 | ||
| GenBank mRNA | BC079031.1 | - | 7 | 1405 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spag8| Gene Symbol: Spag8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173555.1 | NP_001167026.1 | 7 | 1405 | ||
| GenBank mRNA | BC079031.1 | - | 7 | 1405 | ||