siRNA Details
| Gene Symbol: Msantd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108664.1 | NP_001102134.1 | 3 | 645 | ||
| XM_008763701.1 | XP_008761923.1 | 2 | 886 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msantd3| Gene Symbol: Msantd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108664.1 | NP_001102134.1 | 3 | 645 | ||
| XM_008763701.1 | XP_008761923.1 | 2 | 886 | |||