siRNA Details
Gene Symbol: Tmem38b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014191.1 | NP_001014213.1 | 5, 6 | 685 | ||
GenBank mRNA | BC079336.1 | - | 5, 6 | 685 | 7106021 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem38b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014191.1 | NP_001014213.1 | 5, 6 | 685 | ||
GenBank mRNA | BC079336.1 | - | 5, 6 | 685 | 7106021 |