siRNA Details
| Gene Symbol: Fkbp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763840.1 | XP_008762062.1 | 25 | 2950 | ||
| GenBank mRNA | FQ230831.1 | - | 24 | 2841 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fkbp15| Gene Symbol: Fkbp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763840.1 | XP_008762062.1 | 25 | 2950 | ||
| GenBank mRNA | FQ230831.1 | - | 24 | 2841 | ||