siRNA Details
| Gene Symbol: LOC102556352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238232.2 | XP_006238294.1 | 9 | 1435 | ||
| Gene Symbol: Fkbp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763840.1 | XP_008762062.1 | 16 | 1819 | ||
| GenBank mRNA | FQ230831.1 | - | 15 | 1740 | ||

LOC102556352
Genomic Map