siRNA Details
| Gene Symbol: Hacd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108669.1 | NP_001102139.1 | 3 | 226 | ||
| XM_006238376.2 | XP_006238438.1 | 3 | 260 | |||
| XM_008763794.1 | XP_008762016.1 | 3 | 270 | |||
| XM_008763795.1 | XP_008762017.1 | 3 | 269 | |||
| GenBank mRNA | FQ234476.1 | - | 3 | 222 | ||

Hacd4
Genomic Map