siRNA Details
Gene Symbol: Alg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033709.1 | NP_001028881.1 | 13 | 1444 | ||
GenBank mRNA | BC101850.1 | - | 13 | 1444 | 5621707 | |
FQ210318.1 | - | 13 | 1459 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Alg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033709.1 | NP_001028881.1 | 13 | 1444 | ||
GenBank mRNA | BC101850.1 | - | 13 | 1444 | 5621707 | |
FQ210318.1 | - | 13 | 1459 |