siRNA Details
| Gene Symbol: Efcab7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191866.1 | NP_001178795.1 | 12 | 1690 | ||
| XM_008763885.1 | XP_008762107.1 | 12 | 1740 | |||
| XM_008763886.1 | XP_008762108.1 | 12 | 1678 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efcab7| Gene Symbol: Efcab7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191866.1 | NP_001178795.1 | 12 | 1690 | ||
| XM_008763885.1 | XP_008762107.1 | 12 | 1740 | |||
| XM_008763886.1 | XP_008762108.1 | 12 | 1678 | |||