siRNA Details
| Gene Symbol: Zyg11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_342878.8 | XP_342879.3 | 10 | 2111 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zyg11b| Gene Symbol: Zyg11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_342878.8 | XP_342879.3 | 10 | 2111 | ||