siRNA Details
| Gene Symbol: Lrrc41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009710.1 | NP_001009710.1 | 4 | 1497 | ||
| XM_006238682.2 | XP_006238744.1 | 4 | 1882 | |||
| GenBank mRNA | BC087037.1 | - | 4 | 1497 | 7133544 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc41| Gene Symbol: Lrrc41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009710.1 | NP_001009710.1 | 4 | 1497 | ||
| XM_006238682.2 | XP_006238744.1 | 4 | 1882 | |||
| GenBank mRNA | BC087037.1 | - | 4 | 1497 | 7133544 | |