siRNA Details
| Gene Symbol: Scmh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109669.1 | NP_001103139.1 | 10, 11 | 1358 | ||
| XM_008764027.1 | XP_008762249.1 | 10, 11 | 1842 | |||
| XM_008764028.1 | XP_008762250.1 | 10, 11 | 1842 | |||
| XM_008764029.1 | XP_008762251.1 | 9, 10 | 1702 | |||
| XM_008764030.1 | XP_008762252.1 | 9, 10 | 3919 | |||
| XM_008764031.1 | XP_008762253.1 | 10, 11 | 1449 | |||
| XM_008764032.1 | XP_008762254.1 | 8, 9 | 1206 | |||
| GenBank mRNA | BC131847.1 | - | 10, 11 | 1421 | 7100459 | |

Scmh1
Genomic Map