siRNA Details
| Gene Symbol: Scmh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109669.1 | NP_001103139.1 | 14 | 1971 | ||
| XM_008764027.1 | XP_008762249.1 | 14 | 2455 | |||
| XM_008764029.1 | XP_008762251.1 | 13 | 2315 | |||
| XM_008764030.1 | XP_008762252.1 | 13 | 4532 | |||
| XM_008764031.1 | XP_008762253.1 | 14 | 2062 | |||
| XM_008764032.1 | XP_008762254.1 | 12 | 1819 | |||
| GenBank mRNA | BC131847.1 | - | 14 | 2034 | 7100459 | |

Scmh1
Genomic Map