siRNA Details
Gene Symbol: Scmh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109669.1 | NP_001103139.1 | 7 | 731 | ||
XM_008764027.1 | XP_008762249.1 | 7 | 1215 | |||
XM_008764028.1 | XP_008762250.1 | 7 | 1215 | |||
XM_008764029.1 | XP_008762251.1 | 6 | 1075 | |||
XM_008764030.1 | XP_008762252.1 | 6 | 3292 | |||
XM_008764031.1 | XP_008762253.1 | 7 | 822 | |||
XM_008764032.1 | XP_008762254.1 | 5 | 579 | |||
GenBank mRNA | BC131847.1 | - | 7 | 794 | 7100459 |
