siRNA Details
| Gene Symbol: Zmym6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108681.2 | NP_001102151.1 | 15 | 3217 | ||
| XM_006238892.2 | XP_006238954.1 | 14 | 4048 | |||
| XM_006238894.2 | XP_006238956.1 | 15 | 3095 | |||
| GenBank mRNA | FQ221018.1 | - | 15 | 3287 | ||

Zmym6
Genomic Map