siRNA Details
| Gene Symbol: Srsf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025738.1 | NP_001020909.1 | 1, 2 | 169 | ||
| XM_006239220.2 | XP_006239282.1 | 1, 2 | 212 | |||
| XM_006239221.2 | XP_006239283.1 | 1, 2 | 212 | |||
| XM_006239222.2 | XP_006239284.1 | 1, 2 | 210 | |||
| XM_006239223.2 | XP_006239285.1 | 1, 2 | 211 | |||
| GenBank mRNA | BC098831.1 | - | 1, 2 | 169 | 7378554 | |

Srsf10
Genomic Map