siRNA Details
| Gene Symbol: Zbtb40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191871.1 | NP_001178800.1 | 3 | 509 | ||
| XM_006239232.2 | XP_006239294.1 | 3 | 547 | |||
| XM_006239233.2 | XP_006239295.1 | 3 | 547 | |||
| XM_008764263.1 | XP_008762485.1 | 3 | 548 | |||
| XM_008764264.1 | XP_008762486.1 | 3 | 6106 | |||
| XM_008764265.1 | XP_008762487.1 | 4 | 1249 | |||
| XM_008764266.1 | XP_008762488.1 | 3 | 5491 | |||
| XM_008764267.1 | XP_008762489.1 | 4 | 638 | |||
| XM_008764268.1 | XP_008762490.1 | 2 | 441 | |||
| XM_008764269.1 | XP_008762491.1 | 3 | 548 | |||

Zbtb40
Genomic Map