siRNA Details
| Gene Symbol: Pqlc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108689.2 | NP_001102159.1 | 2 | 302 | ||
| XM_006239238.2 | XP_006239300.1 | 2 | 633 | |||
| XM_006239239.2 | XP_006239301.1 | 4 | 1062 | |||
| XM_006239240.2 | XP_006239302.1 | 4 | 462 | |||
| XM_006239244.2 | XP_006239306.1 | 4 | 466 | |||
| GenBank mRNA | BC158607.1 | - | 4 | 415 | 7192223 | |

Pqlc2
Genomic Map