siRNA Details
| Gene Symbol: Mthfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239413.2 | XP_006239475.1 | 6 | 2402 | ||
| XM_006239414.2 | XP_006239476.1 | 6 | 900 | |||
| XM_006239416.1 | XP_006239478.1 | 6 | 1067 | |||
| XM_006239417.1 | XP_006239479.1 | 6 | 1064 | |||
| XM_006239418.1 | XP_006239480.1 | 5 | 999 | |||
| XM_342975.6 | XP_342976.3 | 6 | 2405 | |||
| GenBank mRNA | U57049.1 | - | 5 | 793 | ||

Mthfr
Genomic Map