siRNA Details
| Gene Symbol: Mthfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239413.2 | XP_006239475.1 | 5 | 2228 | ||
| XM_006239414.2 | XP_006239476.1 | 5 | 726 | |||
| XM_006239416.1 | XP_006239478.1 | 5 | 893 | |||
| XM_006239417.1 | XP_006239479.1 | 5 | 890 | |||
| XM_006239418.1 | XP_006239480.1 | 4 | 825 | |||
| XM_342975.6 | XP_342976.3 | 5 | 2231 | |||
| GenBank mRNA | U57049.1 | - | 4 | 619 | ||

Mthfr
Genomic Map