siRNA Details
| Gene Symbol: Heatr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191064.1 | NP_001177993.1 | 27, 28 | 4246 | ||
| GenBank mRNA | BC079335.1 | - | 1, 2 | 117 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Heatr5b| Gene Symbol: Heatr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191064.1 | NP_001177993.1 | 27, 28 | 4246 | ||
| GenBank mRNA | BC079335.1 | - | 1, 2 | 117 | ||