siRNA Details
| Gene Symbol: Gemin6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009466.2 | NP_001009466.1 | 3 | 512 | ||
| XM_006239647.2 | XP_006239709.1 | 3 | 614 | |||
| GenBank mRNA | AY053516.1 | - | 3 | 513 | ||
| BC158565.1 | - | 4 | 718 | |||
| FQ209637.1 | - | 1 | 223 | |||

Gemin6
Genomic Map