siRNA Details
Gene Symbol: Tmem178a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004282.1 | NP_001004282.1 | 2 | 468 | ||
GenBank mRNA | BC079338.1 | - | 2 | 498 | 7106500 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem178a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004282.1 | NP_001004282.1 | 2 | 468 | ||
GenBank mRNA | BC079338.1 | - | 2 | 498 | 7106500 |