siRNA Details
| Gene Symbol: Tmem178a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004282.1 | NP_001004282.1 | 3, 4 | 654 | ||
| GenBank mRNA | BC079338.1 | - | 3, 4 | 684 | 7106500 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem178a| Gene Symbol: Tmem178a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004282.1 | NP_001004282.1 | 3, 4 | 654 | ||
| GenBank mRNA | BC079338.1 | - | 3, 4 | 684 | 7106500 | |