siRNA Details
| Gene Symbol: Ttc7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100756.1 | NP_001094226.1 | 14, 15 | 1894 | ||
| XM_008764464.1 | XP_008762686.1 | 7, 8 | 2963 | |||
| GenBank mRNA | BC168226.1 | - | 14, 15 | 1894 | 7371309 | |

Ttc7a
Genomic Map