siRNA Details
| Gene Symbol: Ppp1r21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271305.1 | NP_001258234.1 | 16, 17 | 1815 | ||
| XM_008764482.1 | XP_008762704.1 | 15, 16 | 1728 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r21| Gene Symbol: Ppp1r21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271305.1 | NP_001258234.1 | 16, 17 | 1815 | ||
| XM_008764482.1 | XP_008762704.1 | 15, 16 | 1728 | |||