siRNA Details
| Gene Symbol: Bre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199270.1 | NP_954891.1 | 2 | 94 | ||
| XM_006239802.2 | XP_006239864.1 | 3 | 203 | |||
| GenBank mRNA | BC061573.1 | - | 2 | 94 | 6887072 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bre| Gene Symbol: Bre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199270.1 | NP_954891.1 | 2 | 94 | ||
| XM_006239802.2 | XP_006239864.1 | 3 | 203 | |||
| GenBank mRNA | BC061573.1 | - | 2 | 94 | 6887072 | |