siRNA Details
| Gene Symbol: Seli | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134754.1 | NP_001128226.2 | 3 | 241 | ||
| GenBank mRNA | BC168235.1 | - | 3 | 241 | 7444430 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Seli| Gene Symbol: Seli | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134754.1 | NP_001128226.2 | 3 | 241 | ||
| GenBank mRNA | BC168235.1 | - | 3 | 241 | 7444430 | |