siRNA Details
| Gene Symbol: Snx13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108708.1 | NP_001102178.1 | 12 | 1454 | ||
| XM_006240022.2 | XP_006240084.1 | 14 | 2110 | |||
| XM_006240023.2 | XP_006240085.1 | 14 | 2108 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx13| Gene Symbol: Snx13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108708.1 | NP_001102178.1 | 12 | 1454 | ||
| XM_006240022.2 | XP_006240084.1 | 14 | 2110 | |||
| XM_006240023.2 | XP_006240085.1 | 14 | 2108 | |||