siRNA Details
| Gene Symbol: Stxbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191872.1 | NP_001178801.1 | 2 | 306 | ||
| XM_006240067.2 | XP_006240129.1 | 2 | 1868 | |||
| XM_008764658.1 | XP_008762880.1 | 3 | 371 | |||
| XM_008764659.1 | XP_008762881.1 | 2 | 323 | |||

Stxbp6
Genomic Map