siRNA Details
| Gene Symbol: Ppp2r3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014196.1 | NP_001014218.1 | 4 | 405 | ||
| XM_008764660.1 | XP_008762882.1 | 3 | 291 | |||
| GenBank mRNA | BC079257.1 | - | 4 | 405 | 7133341 | |
| FQ227548.1 | - | 3 | 291 | |||
| Gene Symbol: LOC102550845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257714.2 | XP_006257776.1 | 2 | 810 | ||

Ppp2r3c
Genomic Map