siRNA Details
| Gene Symbol: Ppp2r3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014196.1 | NP_001014218.1 | 11 | 1126 | ||
| XM_008764660.1 | XP_008762882.1 | 10 | 1012 | |||
| GenBank mRNA | BC079257.1 | - | 11 | 1126 | 7133341 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp2r3c| Gene Symbol: Ppp2r3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014196.1 | NP_001014218.1 | 11 | 1126 | ||
| XM_008764660.1 | XP_008762882.1 | 10 | 1012 | |||
| GenBank mRNA | BC079257.1 | - | 11 | 1126 | 7133341 | |