siRNA Details
Gene Symbol: Pax9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039539.1 | NP_001034628.1 | 2 | 589 | ||
GenBank mRNA | AB248957.1 | - | 2 | 589 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pax9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039539.1 | NP_001034628.1 | 2 | 589 | ||
GenBank mRNA | AB248957.1 | - | 2 | 589 |