siRNA Details
| Gene Symbol: Atp5s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007749.1 | NP_001007750.1 | 2 | 349 | ||
| XM_006240174.2 | XP_006240236.1 | 2 | 252 | |||
| XM_006240175.2 | XP_006240237.1 | 2 | 141 | |||
| XM_006240176.2 | XP_006240238.1 | 2 | 91 | |||
| GenBank mRNA | BC083655.1 | - | 2 | 349 | 7135201 | |

Atp5s
Genomic Map