siRNA Details
| Gene Symbol: Atl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009831.2 | NP_001009831.1 | 4 | 618 | ||
| XM_006240177.1 | XP_006240239.1 | 3 | 554 | |||
| GenBank mRNA | AY581896.2 | - | 4 | 618 | ||
| BC092645.1 | - | 3 | 541 | 7318365 | ||

Atl1
Genomic Map