siRNA Details
| Gene Symbol: Tmx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024800.1 | NP_001019971.1 | 5, 6 | 527 | ||
| XM_006240178.2 | XP_006240240.1 | 5, 6 | 1750 | |||
| GenBank mRNA | BC094308.1 | - | 5, 6 | 527 | 7316392 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmx1| Gene Symbol: Tmx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024800.1 | NP_001019971.1 | 5, 6 | 527 | ||
| XM_006240178.2 | XP_006240240.1 | 5, 6 | 1750 | |||
| GenBank mRNA | BC094308.1 | - | 5, 6 | 527 | 7316392 | |