siRNA Details
| Gene Symbol: Dcaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108716.1 | NP_001102186.1 | 3 | 272 | ||
| XM_006240347.2 | XP_006240409.1 | 3 | 272 | |||
| XM_008764814.1 | XP_008763036.1 | 2 | 320 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcaf4| Gene Symbol: Dcaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108716.1 | NP_001102186.1 | 3 | 272 | ||
| XM_006240347.2 | XP_006240409.1 | 3 | 272 | |||
| XM_008764814.1 | XP_008763036.1 | 2 | 320 | |||