siRNA Details
| Gene Symbol: Angel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108717.1 | NP_001102187.1 | 8 | 1704 | ||
| XM_006240392.2 | XP_006240454.1 | 9 | 2236 | |||
| XM_008764815.1 | XP_008763037.1 | 8 | 1892 | |||
| GenBank mRNA | BC166827.1 | - | 3 | 229 | ||

Angel1
Genomic Map