siRNA Details
| Gene Symbol: RGD1311756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173557.1 | NP_001167028.1 | 9 | 1402 | ||
| XM_006240439.2 | XP_006240501.1 | 10 | 1576 | |||
| XM_006240441.2 | XP_006240503.1 | 10 | 1637 | |||
| XM_006240442.2 | XP_006240504.1 | 8 | 2389 | |||
| XM_008764817.1 | XP_008763039.1 | 10 | 1631 | |||
| GenBank mRNA | FQ221290.1 | - | 8 | 1258 | ||

RGD1311756
Genomic Map