siRNA Details
| Gene Symbol: Vrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012194.1 | NP_001012194.1 | 9 | 1017 | ||
| XM_006240530.2 | XP_006240592.1 | 9 | 843 | |||
| XM_006240531.2 | XP_006240593.1 | 9 | 873 | |||
| XM_006240532.2 | XP_006240594.1 | 9 | 1265 | |||
| XM_006240533.2 | XP_006240595.1 | 9 | 752 | |||
| XM_008764822.1 | XP_008763044.1 | 9 | 1265 | |||
| GenBank mRNA | BC079130.1 | - | 9 | 1017 | 7127355 | |

Vrk1
Genomic Map