siRNA Details
| Gene Symbol: Zfyve21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108725.1 | NP_001102195.1 | 3 | 264 | ||
| XM_006240639.2 | XP_006240701.2 | 3 | 320 | |||
| XM_006240641.2 | XP_006240703.2 | 3 | 320 | |||
| XM_006240642.2 | XP_006240704.2 | 3 | 320 | |||
| XM_006240643.2 | XP_006240705.2 | 3 | 320 | |||
| XM_006240644.2 | XP_006240706.2 | 3 | 320 | |||
| XM_008764946.1 | XP_008763168.1 | 3 | 320 | |||

Zfyve21
Genomic Map