siRNA Details
| Gene Symbol: RGD1307315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008377.1 | NP_001008378.1 | 1 | 383 | ||
| GenBank mRNA | BC086564.1 | - | 1 | 383 | 7305233 | |
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 RGD1307315
RGD1307315| Gene Symbol: RGD1307315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008377.1 | NP_001008378.1 | 1 | 383 | ||
| GenBank mRNA | BC086564.1 | - | 1 | 383 | 7305233 | |
 Back To Top
 Back To Top