siRNA Details
| Gene Symbol: Slc39a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108728.1 | NP_001102198.1 | 2 | 636 | ||
| XM_006240760.2 | XP_006240822.1 | 3 | 541 | |||
| XM_006240761.2 | XP_006240823.1 | 3 | 553 | |||
| XM_006240762.2 | XP_006240824.1 | 4 | 661 | |||
| XM_006240763.2 | XP_006240825.1 | 4 | 649 | |||
| XM_006240764.2 | XP_006240826.1 | 2 | 445 | |||
| XM_006240766.2 | XP_006240828.1 | 3 | 756 | |||
| XM_006240767.2 | XP_006240829.1 | 1 | 64 | |||
| XM_008765027.1 | XP_008763249.1 | 3 | 575 | |||
| XM_008765028.1 | XP_008763250.1 | 3 | 541 | |||

Slc39a5
Genomic Map