siRNA Details
| Gene Symbol: Cdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199501.1 | NP_955795.1 | 6, 7 | 933 | ||
| XM_006240778.1 | XP_006240840.1 | 7, 8 | 1153 | |||
| GenBank mRNA | BC061832.1 | - | 6, 7 | 933 | 5599608 | |
| D28753.1 | - | 7, 8 | 969 | |||

Cdk2
Genomic Map