siRNA Details
| Gene Symbol: Dot1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108733.1 | NP_001102203.1 | 5 | 635 | ||
| XM_006240959.2 | XP_006241021.1 | 5 | 587 | |||
| XM_006240960.2 | XP_006241022.1 | 5 | 587 | |||
| XM_006240961.2 | XP_006241023.1 | 5 | 441 | |||

Dot1l
Genomic Map