siRNA Details
| Gene Symbol: Mier2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108737.1 | NP_001102207.1 | 9 | 836 | ||
| XM_006240977.2 | XP_006241039.1 | 9 | 1071 | |||
| XM_006240978.1 | XP_006241040.1 | 9 | 858 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mier2| Gene Symbol: Mier2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108737.1 | NP_001102207.1 | 9 | 836 | ||
| XM_006240977.2 | XP_006241039.1 | 9 | 1071 | |||
| XM_006240978.1 | XP_006241040.1 | 9 | 858 | |||