siRNA Details
Gene Symbol: Zfp870 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163062.1 | NP_001156534.1 | 4 | 499 | ||
XM_006241082.2 | XP_006241144.1 | 4 | 586 |
Gene Symbol: LOC102548431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765186.1 | XP_008763408.1 | 4 | 1281 |
