siRNA Details
| Gene Symbol: Pram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241132.2 | XP_006241194.1 | 6 | 2066 | ||
| XM_008765204.1 | XP_008763426.1 | 6 | 2066 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pram1| Gene Symbol: Pram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241132.2 | XP_006241194.1 | 6 | 2066 | ||
| XM_008765204.1 | XP_008763426.1 | 6 | 2066 | |||