siRNA Details
| Gene Symbol: Pwp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191877.1 | NP_001178806.1 | 11 | 1204 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pwp1| Gene Symbol: Pwp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191877.1 | NP_001178806.1 | 11 | 1204 | ||