siRNA Details
| Gene Symbol: Appl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108741.1 | NP_001102211.1 | 11 | 977 | ||
| XM_006241201.2 | XP_006241263.1 | 11 | 1018 | |||
| GenBank mRNA | BC168167.1 | - | 11 | 892 | 9021772 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Appl2| Gene Symbol: Appl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108741.1 | NP_001102211.1 | 11 | 977 | ||
| XM_006241201.2 | XP_006241263.1 | 11 | 1018 | |||
| GenBank mRNA | BC168167.1 | - | 11 | 892 | 9021772 | |