siRNA Details
| Gene Symbol: Hsp90b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012197.2 | NP_001012197.2 | 12 | 1633 | ||
| GenBank mRNA | BC081917.1 | - | 12 | 1633 | 7112411 | |
| FQ229309.1 | - | 1 | 47 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsp90b1| Gene Symbol: Hsp90b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012197.2 | NP_001012197.2 | 12 | 1633 | ||
| GenBank mRNA | BC081917.1 | - | 12 | 1633 | 7112411 | |
| FQ229309.1 | - | 1 | 47 | |||